PIG® Outdoor Pan, New Pig
# de catalogue 56620-092
Fournisseur: New Pig
Spécifications
- Description:Outdoor drip pan
- Capacity:Absorbs up to 1/2 gal. per Pan
- Supplier No.:PAN401
Spécifications
A propos de cet article
Absorbs and retains oils and oil-based liquids, including lubricants and fuels, without taking in a drop of water.
- Mesh top lets liquid pass into absorbent filler housed in a chemical-resistant pan
- Rainwater passes through slits in side of pan to avoid puddling
- Low-profile pan fits in tight spaces and won't tip over
Includes PIG Oil-Only Mat to absorb any additional oily discharge and protect the ground.
Customers Who Bought This Also Bought