VWR® Specific Gravity Bottle
# de catalogue 16629-009
Fournisseur: VWR International
Spécifications
- Cap Size:14/12
- Capacity:25 ml (0.85 oz.)
- ST Stopper No.:10/15
Spécifications
A propos de cet article
Specific-gravity bottles are flasks made to hold a known volume of liquid at a specified temperature.
- Borosilicate glass construction
- Not adjusted
The bottle is weighed, filled with the liquid whose specific gravity is to be found, and weighed again.
Weld type pycnometer devised for specific gravity of volatile liquids. [ST] glass stopper and cap reduce evaporation.
Ordering information: Supplied complete with [ST] stopper and [ST] cap.
Customers Who Bought This Also Bought