GUST® Dusters, Stoner®
Fournisseur: Stoner
Certificats
A propos de cet article
GUST® Dusters blast clean, filtered, moisture-free, high-pressue gas to remove dust, dirt, lint, hair, sand, sawdust, moisture, and microscopic debris from hard-to-reach areas or delicate surfaces such as keyboards, printers, computers, cameras, lenses, audio gear, sensitive instruments, and other office equipment or electronics.
- Blast clean, filtered, moisture-free, high-pressure gas to remove dust, hair, and moisture from hard-to-reach areas or delicate surfaces
- A unique variable pressure trigger allows for control of dusting pressure
- Gas is 100% pure, non-flammable, non-ozone depleting, and filtered to 0.2 microns
- Spray Anyway duster features a 360° valve that allows the user to spray from any angle or upside down without frosting
Duster will not harm glass, optics, metals, plastics, rubber, or hardware. A unique variable pressure trigger allows for control of dusting pressure. Duster gas is 100% pure, non-flammable, non-ozone depleting, and filtered to 0.2 microns.
The Spray Anyway duster features a 360° valve that allows the user to spray from any angle or upside down without frosting.
Customers Who Bought This Also Bought