VWR® Dust'it™ Blower
# de catalogue 36934-142
Fournisseur: VWR International
Spécifications
- Dimensions:5.7Dia. x 12.7L cm (2.25x5")
- Volume:57 g (2 oz.)
- Description:VWR® Dust'it™ bloer56.7 g
- Cat. no.:36934-142
Spécifications
A propos de cet article
Providing high-pressure, pinpoint jets of air, this powerful bulb duster forcibly blows away dust, lint, and dirt.
- Forcibly blows away dust, lint, and dirt
- Ideal for delicate, hard-to-clean areas
- Nonconductive plastic sleeve tip prevents scratches to surfaces
- One-way inlet valve at the top keeps air from being drawn in
Ideal for delicate, difficult-to-clean, and inaccessible equipment. The nonconductive plastic sleeve tip prevents scratches for cleaning optical lenses, slides, circuit boards, and cuvets. The one-way inlet valve at the top stops air from being drawn through the tip. Varying the “squeeze” provides for controlled delivery.
Customers Who Bought This Also Bought