Utility Cart, Labconco®
# de catalogue 82006-848
Fournisseur: Labconco
Product available on GSA Advantage®
Spécifications
- Dimensions:347/8x19x363/8"
- Cat. no.:82006-848
- Compartment Dimensions:153/4x147/8x41/4"
- Shipping Weight:44 lbs.
- Load Capacity:400 lbs.
- Supplier No.:8007000
Spécifications
A propos de cet article
Features two open shelves for the transport and support of small electrical equipment
Carts have a 2.5cm (1") tubular steel frame with a corrosion-resistant dry powder glacier white epoxy coating. Carts feature four 10.2cm (4") diameter casters (two with toe locks) and two handles with ergonomic extruded rubber grips. Carts include two corrosion-resistant, one-piece, vacuum-formed ABS plastic drawers to hold small labware and supplies, and a six-outlet 115V, 60Hz, 12A power strip with a cord bracket and a three-wire cord with plug. Supports up to 181kg (400lbs.).
May also be used as a compact workstation for laboratory equipment.
Ordering information: Manufacturer's lifetime warranty. Shipped fully assembled.
Customers Who Bought This Also Bought