VWR® Utility Carts, Stainless Steel
New Product
These durable utility carts feature a unitized, electronically welded stainless steel angled leg design.
- Unitized, electronically welded stainless steel angled leg design
- Cleans and sanitizes easily
- Three shelves
Sound deadening panels under shelves restrict vibration and noise
Note: Capacity is the total cart capacity across all of the shelves with the weight evenly dispersed.
Bumpers on legs and handles protect walls and furniture.
Frequently Bought Together