Cytology Spin Slides, Azer Scientific
Fournisseur: Azer Scientific Inc.
Certificats
A propos de cet article
Cytology Spin slides are compatible with Thermo Shandon centrifuge systems and are designed to aid in the quick and easy location of the centrifuged smear.
- Thermo Shandon centrifuge system-compatible
- Single or double wells available
- Adhesive coating available
- Well placement in the same position of centrifuge specimen deposit
Available with single or double wells.
Slides feature well placement in the exact position where the centrifuge specimen is deposited. Available with or without adhesive coating.
Packaging: 72 slides per box, two boxes per case
Customers Who Bought This Also Bought