AzerMark™ Etching Slides, Azer Scientific
# de catalogue 89087-558
Fournisseur: Azer Scientific Inc.
Minority or Woman-Owned Business Enterprise
Spécifications
- Description:Azermark™ etching slides
- L×W:75×25 mm (3×1")
- Thickness:1.0 mm
- Cat. no.:89087-558
- Supplier No.:EMS300BW
Spécifications
A propos de cet article
These slides are designed for use with automated slide etch labeling machines.
The Diamond™ White Glass Beveled Edge Microscope Slides are offered in bright, attractive colors. They feature Beveled Edges with Clipped Corners and are pre-cleaned and supplied ready for use.
- White glass with ground edges
- Black and white labeling area
- Dimensions: 75 x 25 x 1 mm
- Adhesion available
These slides are designed for use with automated slide etch labeling machines.
The slides have white glass with ground edges, and the frosted coating retains most laboratory marking methods.
They are available with or without adhesion and include a black and white labeling area.
Ordering information: Slides are packaged 72 per box, 20 boxes per case (10 gross).
Customers Who Bought This Also Bought