Colorfrost® Plus Microscope Slides, Erie Scientific (Thermo Scientific)
Fournisseur: Erie Scientific
Certificats
A propos de cet article
A permanent positive charge applied to the slide surface allows slides to hold tissue sections without the need for expensive or time-consuming preparatory steps in the laboratory
A permanent positive charge applied to the slide surface allows slides to hold tissue sections without the need for expensive or time-consuming preparatory steps in the laboratory
This treatment virtually eliminates tissue loss during staining and electrostatically attracts frozen tissue sections and cytology preparations, binding them to the slide. Color-coded for quick and simple identification.
Customers Who Bought This Also Bought