Superfrost® Cytogenetics Slides, Erie Scientific (Thermo Scientific)
# de catalogue 89050-838
Fournisseur: Erie Scientific
Spécifications
- Description:Frosted slides, Superfrost
- L×W:75×25 mm (3×1")
- Thickness:1.0 mm
- Cat. no.:89050-838
- Supplier No.:4951-7140
Spécifications
A propos de cet article
The consisetency and quality of these slides allows for reliable chromosome spreads, while the hydrophilic surface improves retention of metaphase cells
The consisetency and quality of these slides allows for reliable chromosome spreads, while the hydrophilic surface improves retention of metaphase cells
Slides have a Superfrost® white tab for permanent marking.
Customers Who Bought This Also Bought