Clipped Corner Microscope Slides, Epredia™
Fournisseur: Erie Scientific
Certificats
A propos de cet article
Slides feature consistent 45° clipped corners for ease of handling and to aid with manual blood smears
Offering exceptional cleanliness and resistance to sticking, these slides are ideal for technicians who routinely prepare large numbers of slides. Superfrost® slides have an opaque coating that takes and retains most laboratory marking methods better than plain or conventional frosted slides. The Superfrost® coating is impervious to most common laboratory chemicals and reagents.
Customers Who Bought This Also Bought