KONTES® Four-Neck Reaction Vessel Lids, Kimble Chase
Fournisseur: DWK Life Sciences (KIMBLE)
Certificats
A propos de cet article
For use with cylindrical or spherical reaction vessels with a 10.2cm (4") I.D
flange. Available with two standard side necks and one thermometer neck or three standard side necks. Thermometer neck [ST] joint: 10/30.
Ordering information: For clamps, additional flange sizes, and additional [ST] joint numbers, contact your Avantor sales representative.
Customers Who Bought This Also Bought