KONTES® Three-Neck Reaction Vessel Lids, Kimble Chase
Fournisseur: DWK Life Sciences (KIMBLE)
Certificats
A propos de cet article
For use with cylindrical or spherical reaction flasks with a 10.2cm (4") I.D
flange.
Ordering information: For clamps and other reaction vessel lids, contact your Avantor sales representative.
Customers Who Bought This Also Bought