Four-Neck Reaction Vessel Lids, Chemglass
Fournisseur: Chemglass
Certificats
A propos de cet article
These four neck reaction vessel lids have a flat ground surface allowing it to be greased or used with an o-ring.
Lids with thermometer side neck have a 10/30 outer joint which will permit a thermometer to be inserted vertically to the bottom of the vessel.
Ordering information: Four neck lids are available with joint sizes and configurations other than those listed.
Customers Who Bought This Also Bought