Corning® Frosted Micro Slides, Corning
Fournisseur: Corning
Certificats
A propos de cet article
For use in nonsterile cytology and pathology applications.
Made of water-white glass for maximum clarity. Precleaned and ready for use. Inspected for chips and rough edges before packaging. Slides are sandblasted and not etched, resulting in high legibility against most laboratory bench backgrounds.
Ordering information: Packaged 1/2 gross per box. Slides are packed vertically for easy lift out.
Customers Who Bought This Also Bought