SP Wilmad-LabGlass Four-Neck Reaction Vessel Lids, O-Ring Flange, SP Industries
Fournisseur: Avantor
Certificats
A propos de cet article
Vessel lids feature one center neck and three side necks 120° apart
All four necks have [ST] joints. Lids with O-ring size 348 fit reaction vessels 14204-802 and -874; lids with O-ring size 359 fit vessels 14204-050, -790, and 14203-984.
Customers Who Bought This Also Bought