SP Wilmad-LabGlass Four-Neck Reaction Vessel Covers, SP Industries
Fournisseur: SP Industries
Certificats
A propos de cet article
Featuring a center neck and three side necks with full length outer Standard Taper joints that are 120° apart, these borosilicate glass reaction vessel lids have a conical flange
Bottom flange ID is 4" and ground for a leak-proof fit with a reaction flask. For use with reaction vessels LG-8010 through LG-8026
Customers Who Bought This Also Bought