Secondary Spill Containment Pallets with Grating, Eagle Mfg
Fournisseur: Eagle Manufacturing
Certificats
A propos de cet article
Pallets provide safe support and spill containment for 208.2 L (55 gal.) drums.
Corrosion-resistant pallets consist of a translucent tub with open grating to permit quick inspection of possible spills. Grating has 4.4 cm (1³/₄") square openings and is removable for easy cleaning. Transportable by pallet jack or forklift. Pallet units have a 2 cm (³/₄") drain with plug to facilitate cleaning of basin.
Certifications: Meets EPA requirements for secondary spill containment of hazardous materials 40 CFR 264.175.
Customers Who Bought This Also Bought