PIG® Economy Poly Spill Containment Pallet, New Pig
Fournisseur: New Pig
Certificats
A propos de cet article
Polyethylene pallet supports four drums.
- Large sump contains leaks, can be emptied via drain bilge
- Four way forklift entry
- Sturdy poly grates won't corrode or rust
Economical pallets are strong enough to handle long-term storage of fully loaded steel drums. Low-density polyethylene (LDPE) construction resists UV rays, rust, corrosion and most chemicals. Textured grating adds traction and lifts out for easy sump access.
Available with or without drain.
Customers Who Bought This Also Bought