A propos de cet article
SGE autosampler syringes feature Diamond Syringe Technology
Color coded by volume for easy identification of installed syringes. Packaged in 100% recyclable materials.
Enhancements to SGE Diamond syringes result in improved solvent resistance, great temperature ranges,and increased operational smoothness. These improvements deliver longer syringe life and an improved cycle life of up to ten times that of a similar syringes. Syringes offer a new level of accuracy due to reduced carryover and no risk of adhesive contamination.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Catalog No:
- 60361-014
- 60362-186
- 60362-188
- 60362-622
- 60361-132
- Point style:
- Cone
- Cone
- Cone
- Cone
- Cone