Flathead Stoppers, Color-Coded, [ST] Bottle and Apparatus Style, Kimble Chase
Fournisseur: DWK Life Sciences (KIMBLE)
Certificats
A propos de cet article
Use these flathead PTFE stoppers with bottles and ground jointed ware
Top of stopper has color button insert for color coding.
Customers Who Bought This Also Bought