Adhesive Microscope Slide Wells, Diversified Biotech
Fournisseur: Diversified Biotech
Certificats
A propos de cet article
Disposable liquid-holding wells are ideal for examining or counting tissue culture cells, yeast, bacteria, and other microrganisms.
These precut, twin adhesive well stickers firmly adhere to microscope slides. Wells are chemically inert and made of polyester with an acrylic adhesive. Compatible with different styles of microscopes.
Diameter: 1 cm (3/8")
Customers Who Bought This Also Bought