ContactUsLinkComponent
Gastight® 1000 Series SampleLock™ Purge and Trap Syringe, Hamilton
Gastight® 1000 Series SampleLock™ Purge and Trap Syringe, Hamilton
# de catalogue 60373-630
Fournisseur:  Hamilton
Purge and Trap Syringe
Gastight® 1000 Series SampleLock™ Purge and Trap Syringe, Hamilton
# de catalogue 60373-630
Fournisseur:  Hamilton
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Volume:
    5000 µl (5 ml)
  • Description:
    Model 1005 Purge and Trap Syringe
  • Supplier No.:
    81570
Spécifications
A propos de cet article
Purge and Trap syringe allows easy removal of the syringe plunger for loading water samples into the syringe barrel
Designed for the analysis of drinking water samples according to EPA purge and trap concentration techniques (EPA methods 502.1, 502.2, 503.1, 524.1 and 524.2).
Gastight® syringes are ideal for dispensing both liquids and gases. They have a precision-machined PTFE plunger tip which creates a leak-free seal. With the tight fit, the tip essentially wipes the interior of the syringe barrel free of sample. This feature is particularly useful with heterogeneous samples as it reduces the chance that a deposit will occur and cause the plunger to freeze.
An internal standard or surrogate sample can be added directly to the sample through the twist valve with a 10μL syringe. The sample syringe can also be connected directly to the purge and trap valve via the integral male luer fitting. Fluid path includes SST, borosilicate glass, and PTFE. Autoclavable. Gas sterilizable
Ordering information: Needle is not included; contact your Avantor representative for more information.
Customers Who Bought This Also Bought