TexWrite® 22 Heat-Resistant Cleanroom Bond Paper, Texwipe®
Supplier: Texwipe
Certificates
About this item
Multi-purpose cellulose paper is saturated with synthetic latex to reduce the risk of particle generation.
- Features ultra-low levels of particulates, extractables, and ions
- Use in all high-speed laser copiers and printers and standard copier systems
- Dimensionally stable to prevent jamming in copiers and printers
- Double shrink-wrapped
Basis weight: 80 g/m²
Ions: 85 ppm Sodium, 50 ppm Chloride
Opacity: 74%
Particle generation (>0,5 µm) - Texwipe wet test No. 5: 4.8 million/m²
Tear strength, elmendorf tear test: 78 g machine direction, 79 g cross direction
Tensile strength, Fed. Standard No. 191A: 5.3 kg machine direction, 4.5 kg cross direction
W×L: 16x279 mm (8.5x11")
Customers Who Bought This Also Bought