Barnstead/Thermolyne Locator® Cryobiological Storage Systems, Thermo Scientific
Fournisseur: Thermo Fisher Scientific
Certificats
A propos de cet article
Compact cryogenic storage systems feature durable construction and advanced vacuum insulated to minimize liquid nitrogen (LN2) evaporation.
- Vessels feature a secure locking clasp to prevent unauthorized access
- Alarm alerts operator in the event of excessive liquid nitrogen loss
- Systems are suitable for manual and computerized inventory methods
Temperature uniformity ensures that samples are stored at –180 °C (–292 °F) even when the vessel contains less than 5 cm (2") of LN2 and also vacuum insulation reduces the operating costs.
The rugged, steel, 4-wheel storage vessel transporter cart reduces the risk of accidental spillage or tipping of vessels. The racks can accommodate 133×133 mm (5¼×5¼") boxes, which are sold separately.
Ordering information: Liquid nitrogen level monitor is sold separately. Boxes not included. Locator systems are supplied with manufacturer's three-year warranty and stainless steel storage racks.
Spécifications
- Catalog No:
- 89209-048
- 97055-654
- 97055-656
- 55709-800
- 55709-998
- 89209-050
- Model:
- Locator Jr.
- Locator Jr.
- Locator 4
- Locator 4
- Locator Jr.
- Locator 4
- Evaporation rate:
- 0.85 L/day
- 0.85 L/day
- 0.99 L/day
- 0.99 L/day
- 0.85 L/day
- 0.99 L/day
- Ø int.:
- 55.8 cm (22")
- 55.8 cm (22")
- 55.8 cm (22")
- 55.8 cm (22")
- 55.8 cm (22")
- 55.8 cm (22")
- Liquid nitrogen capacity:
- 60 L
- 60 L
- 111 L
- 111 L
- 60 L
- 111 L
- Neck int.Ø:
- 21.5 cm (81/2")
- 21.5 cm (81/2")
- 21.5 cm (81/2")
- 21.5 cm (81/2")
- 21.5 cm (81/2")
- 21.5 cm (81/2")
- Vial capacity:
- 1600
- 1600
- 3600
- 3600
- 1600
- 3600
- Weight Empty:
- 24.6 kg (54.1 lbs.)
- 24.6 kg (54.1 lbs.)
- 35.4 kg (77.8 lbs.)
- 35.4 kg (77.8 lbs.)
- 24.6 kg (54.1 lbs.)
- 35.4 kg (77.8 lbs.)