Drum Tray, Eagle Manufacturing
Fournisseur: Eagle Manufacturing
Certificats
A propos de cet article
Tray with reservoir keeps container away from spills
Ideal for use under drums during dispensing/filling operations. Holds up to 34L (9gal.) of spills when used with a 208.2L (55gal.) drum. Constructed of high-density polyethylene. Use drum tray dolly to easily transport drums between work areas without compromising spill protection.
Customers Who Bought This Also Bought