Polysine™ Adhesion Microscope Slides, Epredia™
# de catalogue 48382-117
Fournisseur: Erie Scientific
Spécifications
- Description:Microscope adhesion slides
- Supplier No.:P4981
Spécifications
A propos de cet article
Slides provide superior cell and tissue adhesion due to a permanent bio-adhesive material that electrostatically and chemically attracts tissues and tissue sections
These slides are particularly effective in applications involving frozen and paraffin embedded sections, cytocentrifuge preparations, and cytology smears. Not affected by chemicals, enzyme predigestion, or heating. Size: 25x75mm.
Customers Who Bought This Also Bought