A propos de cet article
These syringes allow the collection, storage, transport, and analysis of liquid or gaseous samples without the danger of evaporative loss or environmental contamination
The sample lock termination incorporates an on/off syringe valve with removable needle. This termination is used for headspace, environmental sample collection and storage, pre-pressurization of gaseous samples for GC analysis, and sample spiking.
Point style 2 is a sharp, beveled, curved, non-coring needle point recommended for septum penetration.
Optional male luer and female luer lock adapters (sold separately) threads onto the SampleLock™ valve, making the syringes compatible with a multitude of connectors and fittings.
An easy-to-use twist valve and a positive rear stop prevents loss of sample and plunger blowout. Fluid path includes SST, borosilicate glass, and PTFE. Not autoclavable. Gas sterilizable.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Catalog No:
- 60373-602
- 60373-604
- 60373-606
- 60373-608
- 60373-620
- 60373-618
- 60373-600
- 60373-610
- 60373-612
- 60373-614
- 60373-616
- Min. temperature:
- 10°C (50°F)
- 10°C (50°F)
- 10°C (50°F)
- 10°C (50°F)
- 10°C (50°F)
- 10°C (50°F)
- 10°C (50°F)
- 10°C (50°F)
- 10°C (50°F)
- 10°C (50°F)
- 10°C (50°F)
- Needle length:
- 51 mm (2")
- 51 mm (2")
- 51 mm (2")
- 51 mm (2")
- 51 mm (2")
- 51 mm (2")
- 51 mm (2")
- 51 mm (2")
- 51 mm (2")
- 51 mm (2")
- 51 mm (2")
- Point style:
- 2
- 2
- 2
- 2
- 2
- 2
- 2
- 2
- 2
- 2
- 2