Slide Grip, Peel-A-Way® Plastic, Polysciences
# de catalogue 48440-003
Fournisseur: Polysciences Inc
Small Business Enterprise
Spécifications
- Description:Slide Grip, Peel-A-Way® Plastic
- Cat. no.:48440-003
- Supplier No.:19801A-1
Spécifications
A propos de cet article
Slide grip is ideal for staining procedures
It is molded from a special plastic compound which tolerates staining chemicals and solvents, even special metal stains. Fits all Coplin and round-mouth staining jars. Holds up to five slides. Withstands 80°C (176°F) temperatures, to eliminate removing slides for drying.
Customers Who Bought This Also Bought