A propos de cet article
These inert syringes are ideal for liquid and gas samples
Syringes are constructed from borosilicate glass with a fluoropolymer resin-tipped plunger, and assembled with an interference fit between plunger tip and barrel to effect gas sealing properties. Plungers are interchangeable on syringes of the same volume. Luer-lock fittings are not slotted.
These inert syringes are ideal for liquid and gas samples
Syringes are constructed from borosilicate glass with a fluoropolymer resin-tipped plunger, and assembled with an interference fit between plunger tip and barrel to effect gas sealing properties. Plungers are interchangeable on syringes of the same volume. Luer-lock fittings are not slotted.
Syringes are constructed from borosilicate glass with a fluoropolymer resin-tipped plunger, and assembled with an interference fit between plunger tip and barrel to effect gas sealing properties. Plungers are interchangeable on syringes of the same volume. Luer-lock fittings are not slotted.
Note: Syringes can be autoclaved but the plunger should be removed and heated and cooled slowly.
Suitable for manual and dispensing/diluting applications. Dispensed volume is accurate to ±1%.