PYREX® Chromatographic Reagent Atomizers, Corning
Fournisseur: Corning
Certificats
A propos de cet article
Made from Type I Class A - low-expansion borosilicate glass, per ASTM E438 (standard specification for glasses in laboratory apparatus).
They have a full length 24/40 ST joint between the reservoir and spraying device.
Spécifications
- Catalog No:
- 26307-056
- 26307-054
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- I-RECs, ISO 14001 - Monterrey
- I-RECs, ISO 14001 - Monterrey