Ten-Plate Drying Rack, General Glass Blowing
# de catalogue 21432-400
Fournisseur: General Glass Blowing
Spécifications
- Description:Ten-Plate Drying Rack
- Cat. no.:21432-400
- Supplier No.:90-1
Spécifications
A propos de cet article
Carry and store TLC plates without damaging delicate media.
Holds up to 10 standard plates 20x20cm (8x8") or equivalent number of narrower plates. Racked plates may be air- or oven-dried. Plates may be horizontal or vertical. Also store dried layers that are to be used promptly.
Rack is 20-gauge stainless steel with carrying handle. Dimensions: 21Lx20Dx12Hcm (81/4Lx77/8Dx43/4"). Weight: 1.6kg (3.5lbs.).
Customers Who Bought This Also Bought