About this item
These needles are ideal for use in routine blood collection procedures.
- Color-coded for simple identification of needle gauge
- Designed for single use
- Single-sample needles for veterinary use only
They are designed exclusively for single use, and feature color-coded protective needle caps to simplify identification of the needle gauge. Pink caps correspond to 18G needles, yellow caps to 20G, green caps to 21G, and black caps to 22G. Available in two lengths, 25.4 or 38.1 mm (1 or 11/2"). Single-sample needles are designed for veterinary use only.