Monoject™ Blunt Cannulas, Covidien
Fournisseur: Covidien
CE Compliant
Certificats
A propos de cet article
These sterile, stainless steel cannulas feature an aluminum Luer-lock hub and a color-coded needle sheath with a charcoal gray cap
Autoclavable in rigid pack for use in O.R./procedure packs. Latex-free.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.