Synthware Cannula, Stainless Steel, Kemtech America
Supplier: Kemtech America
Certificates
About this item
Designed for the transfer and handling of air sensitive materials.
- Stainless steel construction
- 16, 18, 20 and 22 guage available
- Various lengths available
Customers Who Bought This Also Bought