PTFE Key-Head [ST] Stoppers, Flask Length Joint, Kimble Chase
Fournisseur: DWK Life Sciences (KIMBLE)
Certificats
A propos de cet article
Stoppers are for use with flasks, mixing cylinders, separatory funnels, and more
Designed from ASTM Specification E675.
Color-coded handles indicate stopper size. Stoppers size 9 through 22 are solid. Sizes 32 and 38 are hollow.
Customers Who Bought This Also Bought