Socorex™ Dosys™ 162/172 Basic Self-Refilling Syringes, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
Certificates
About this item
Automatic fill-by-fill syringes feature a fatigue-free two-ring or pistol-grip handle system and interchangeable ground glass barrel
Supplied without a valve system. Autoclavable.
Ordering information: One-year manufacturer's warranty. Supplied with PVC protective sleeve and operating instructions. For luer lock valve assembly parts kit, contact your Avantor representative. For other accessories and replacement parts, see 89044-552 series.
Frequently Bought Together