Brass Cork Borer Sets, Boekel Scientific
# de catalogue 23457-060
Fournisseur: Boekel
Spécifications
- Hole Diameter:5 - 13 mm (³⁄₁₆ - ½")
- Description:Cork borer set, No. 1- 9
- Supplier No.:1601AC
Spécifications
A propos de cet article
Made of brass tubing with strong permanent handles
The cork borer sharpener consists of a solid brass cone with a curved, steel knife blade securely mounted in a wooden handle. It is constructed so that the cutting edge of the borer is trimmed to the correct angle regardless of the borer size. Total length: 19cm (71/2"). For borers with tubes up to 25.4mm (1") O.D.
Cork borer set consists of sequentially numbered, nested tubes that start at 4.5mm (3/16") O.D. and gradually increase in size. A steel punch is included with each set.
Customers Who Bought This Also Bought