Round Developing Chamber, General Glass Blowing
# de catalogue 21432-863
Fournisseur: General Glass Blowing
Spécifications
- Description:Round developing chamber
- Cat. no.:21432-863
- Diameter ×H:12.9×23 cm (5×9")
- Supplier No.:30-1
Spécifications
A propos de cet article
A diameter borosilicate glass jar with flat bottom and lip
Comes with snap-on plastic cap. Holds two 10 x 20cm plates. Uses 75mL of solvent for atmosphere saturation.
Customers Who Bought This Also Bought