Round Developing Chamber, General Glass Blowing | Avantor
Round Developing Chamber, General Glass Blowing
Catalog #
Round Developing Chamber, General Glass Blowing
Catalog #
Supplier:  
Round Developing Chamber, General Glass Blowing
Round Developing Chamber, General Glass Blowing
Product Details & Documents
For 2" wide plates
The borosilicate glass jar has a flat bottom, round lip, and plastic snap-on cap. Uses 25ml of solvent for atmosphere saturation. Handles two 2" wide plates simultaneously. Frequently used to store a series of test solvent systems for selecting the optimum solvent for separating unknown fractions.