About this item
Vitrified silica-cast pestle tips provide enhanced grinding action for difficult and fibrous samples such as muscle or tissue with bone fragments.
- Silica-cast pestle tips
- Sterile
- Closed system prevents aerosol exposure
Unique closed system prevents exposure to aerosols and contact with sample.