Bookshelf Tanks, Scientific Plastics
Fournisseur: Scientific Plastics
Certificats
A propos de cet article
Designed for neutralization and dilution procedures in fume hood cabinets or any area where space is a concern
These seamless tanks are one-piece rotationally molded from virgin high-density polyethylene. Tanks are equipped with a 3.8cm (11/2") male inlet and outlet and a 7.6cm (3") clean-out cap on top.
Customers Who Bought This Also Bought