Dilution Tanks/Traps, Scientific Plastics
Fournisseur: Scientific Plastics
Certificats
A propos de cet article
Lightweight, seamless tanks can be used with water for dilution or with limestone chips for neutralization
Tanks meet ASTM D1248 standards for polyolefin materials.
They are rotationally molded from virgin high-density polyethylene and may be used for continuous service up to 82°C (180°F), or up to 100°C (212°F) in intermittent use. All fittings are made of polyethylene and are welded into all penetrations of tanks.
Customers Who Bought This Also Bought