Cylindrical Tank, Flat-Bottom, PVDF, Saint-Gobain Performance Plastics
# de catalogue 14226-870
Fournisseur: Saint Gobain Performance Plastics
Spécifications
- Volume:56.8 L (15 gal.)
- Description:Cylindrical Tank, Flat Bottom, PVDF
- Cat. no.:14226-870
- O.D.:33 cm (13")
- Depth:68.6 cm (27")
- Graduations:4 L (1 gal.)
- Supplier No.:11500-0015
Spécifications
A propos de cet article
Translucent tank with molded-in graduations
- 2.4 mm (3/32") Wall thickness
Constructed of polyvinylidene fluoride (PVDF) that features low extractables, excellent chemical resistance, and inherent UV resistance. Tank has a maximum service temperature of 110°C (230°F). Tank includes a matching cover with the same thickness as the wall.
Customers Who Bought This Also Bought