Nalgene® Rectangular Tanks, Polypropylene, Thermo Scientific
Fournisseur: Thermo Fisher Scientific
Certificats
A propos de cet article
Tanks have stepped rim for strength and overlapping skirted cover
Polypropylene tank is molded in one piece and is autoclavable
Certifications: Plastic meets USP VI and FDA 21 CFR 177.1520 standards.
Ordering information: Tanks can be modified with factory-installed spigots; contact your Avantor sales representative for more information.
Customers Who Bought This Also Bought