VWR® Contour™ Apron Rollout Keyboard Shelves
Fournisseur: VWR International
Certificats
A propos de cet article
Rollout keyboard shelves feature a low-profile design and come in a range of lengths.
- Smooth, easy-to-maintain surfaces
- Phosphate-coated with a baked, chemical-resistant, powder-coat finish
- Manufactured of prime grade, cold-rolled, U.S. Standard sheet steel with no ragged edges
- Color: light neutral
These shelves feature smooth surfaces with coatings to resist against chemicals.
Customers Who Bought This Also Bought