AMCO AEPA-1® Linear Calibration and Instrument Quality Check Kits, GFS Chemicals
Fournisseur: GFS Chemicals
Certificats
A propos de cet article
Meets EPA linear calibration recommendations
Formazin kits should be used within 30 days of purchase.
Available in a variety of ranges. Each kit includes a 125mL bottle of each stated NTU value.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.