VWR® General Purpose PE Tape
Fournisseur: VWR International
Certificats
A propos de cet article
This high-tack tape has a synthetic rubber-based adhesive that easily bonds to different surfaces such as plastic, concrete, and stainless steel.
- Polyethylene fabrication
- No dead stretch limitations
Tape conforms well and does not have the same “dead stretch” limitations common in other polyethylene tapes. Ideal for polyethylene splicing and surface protection, and as a general replacement in applications where a more aggressive bond is needed.
Customers Who Bought This Also Bought