VWR® Tape and Label Dispenser
Tape and Label Dispenser
Supplier: VWR International
Jump to:
Product Details & Documents
This durable plastic handheld dispenser for tapes and labels accommodates a maximum width of 2.9 cm (11/8").
- Durable plastic construction
- Locking clip
- Dispenses a range of label sizes
Easily dispenses 12.7m (41.7') long rolls of tape in widths of 12.7 mm (1/2"), 19 mm (3/4"), or 25.4 mm (1"), on a 25.4 mm (1") core. Includes two spacers and a plastic locking clip.
Ordering information: Tape and labels sold separately.
Documents
Certificates
Recherche par numéro de lotSpécifications
- Désignation:Tape and Label Dispenser
- Cat. No.:89098-146