Spécifications
- Depth:24"
- Width:2"
- Description:Knee space drawer
- Height:4"
- Material:Plastic laminate
- Cat. no.:33501-200
- Supplier No.:B4740042424
Spécifications
A propos de cet article
Case Systems plastic laminate furniture combines modular and fixed laminate casework with coordinating hardware, trim, countertops, fixtures and many complementary accessories.
- Color: beige
- They are as attractive as they are durable
- Case cabinetry that are adaptability necessary in today’s laboratories
- The key component of casework is the plastic laminate
A perfect choice for laboratory environments, plastic laminate provides superior resistance to chemicals, scratching, and abrasion. The radiused 3 mm PVC edging on doors and drawers further protects the casework from dents and marring while eliminating sharp edges and corners.
The high-quality laboratory grade plastic laminate cabinets are manufactured to exact tolerances with consistent quality standards. Construction features an industrial-grade, environmentally responsible WOODSTALK® engineered board core from Dow BioProducts, using formaldehyde-free resin to bind wheat-straw fiber, and heavy-duty institutional grade hardware.
The unique hardware system provides rigid support for vertical integration without limiting adjustability. Vertical supports are slotted, allowing infinite height adjustment of overhead cabinets, electrical raceways, services, and reagent shelves.